Learn More
Medchemexpress LLC 3',4'-Dimethoxy-alpha-naphthoflavone | 14756-24-2 | MFCD00143008 | 98.5% | 332.35 g·mol^-1 | C21H16O4 | 10 MG
SDP
Catalog No. 5000247702
Encompass_Preferred
DiMNF (3',4'-Dimethoxy-alpha-naphthoflavone) is a small-molecule aryl hydrocarbon receptor (AHR) modulator supplied for research use. It is offered as a solid and as concentrated DMSO solutions for in vitro biochemical and cell-based assays involving AHR signaling and CYP activity.
  • Selective aryl hydrocarbon receptor (AHR) modulator for research applications.
  • High purity (≈98.5%) to support reproducible experimental results.
  • Molecular formula C21H16O4 and molecular weight 332.35 g·mol^-1.
  • Available in multiple pack sizes, including small solid quantities and DMSO solutions.
  • Supplied as a solid with optional preprepared DMSO solutions for convenient dosing.
Catalog No. 50-002-47702 Supplier Medchemexpress LLC Supplier No. HY10322210MG
DiMNF (3',4'-Dimethoxy-alpha-naphthoflavone) is a small-molecule aryl hydrocarbon receptor (AHR) modulator supplied for research use. It is offered as a solid and as concentrated DMSO solutions for in vitro biochemical and cell-based assays involving AHR signaling and CYP activity.
  • Selective aryl hydrocarbon receptor (AHR) modulator for research applications.
  • High purity (≈98.5%) to support reproducible experimental results.
  • Molecular formula C21H16O4 and molecular weight 332.35 g·mol^-1.
  • Available in multiple pack sizes, including small solid quantities and DMSO solutions.
  • Supplied as a solid with optional preprepared DMSO solutions for convenient dosing.