Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-dimethoxy-N-[4-(3-nitrophenyl)-1,3-thiazol-2-yl]benzenesulfonamide | 199666-03-0 | MFCD11040807 | 99.8% | 421.45 | C17H15N3O6S2 | 50MG
SDP

Catalog No. 5000238455
Encompass_Preferred

Ro 61-8048 is a potent, selective inhibitor of kynurenine 3-hydroxylase (kynurenine 3-monooxygenase, KMO) used in biochemical and in vivo research to modulate the kynurenine pathway and elevate extracellular kynurenic acid. It is supplied as a solid research chemical with high reported purity suitable for in vitro and animal studies.

  • Potent KMO inhibitor (IC50 = 37 nM).
  • Selective mechanism targeting kynurenine 3-hydroxylase.
  • High purity: 99.8% (reported by supplier).
  • Molecular weight 421.45 Da; formula C17H15N3O6S2.
  • Physical form: solid; available in small research pack sizes including 50 mg.
  • Suitable for biochemical assays, cellular studies, and in vivo experiments that investigate kynurenine pathway modulation.

Catalog No. 50-002-38455 Supplier Medchemexpress LLC Supplier No. HY1234750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ro 61-8048 is a potent, selective inhibitor of kynurenine 3-hydroxylase (kynurenine 3-monooxygenase, KMO) used in biochemical and in vivo research to modulate the kynurenine pathway and elevate extracellular kynurenic acid. It is supplied as a solid research chemical with high reported purity suitable for in vitro and animal studies.

  • Potent KMO inhibitor (IC50 = 37 nM).
  • Selective mechanism targeting kynurenine 3-hydroxylase.
  • High purity: 99.8% (reported by supplier).
  • Molecular weight 421.45 Da; formula C17H15N3O6S2.
  • Physical form: solid; available in small research pack sizes including 50 mg.
  • Suitable for biochemical assays, cellular studies, and in vivo experiments that investigate kynurenine pathway modulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.