Learn More
Medchemexpress LLC 3,4-dimethoxy-N-[4-(3-nitrophenyl)-1,3-thiazol-2-yl]benzenesulfonamide | 199666-03-0 | MFCD11040807 | 99.8% | 421.45 g/mol | C17H15N3O6S2 | 10MM 1ML
SDP
Catalog No. 5000236415
Encompass_Preferred
Ro 61-8048 is a selective, orally active inhibitor of kynurenine 3-hydroxylase (KMO) used in biochemical and neurological research to modulate the kynurenine pathway. It has a molecular formula of C17H15N3O6S2 and a molecular weight of approximately 421.5 g/mol, and is supplied as a solid or as a 10 mM solution in DMSO for in vitro use.
  • Selective KMO inhibitor with reported IC50 ≈ 37 nM.
  • High purity suitable for research applications.
  • Available as solid (multiple weights) and as a 10 mM solution in DMSO.
  • Off-white to light yellow solid physical form.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
  • Suitable for studies of the kynurenine pathway and neurological disease models.
Catalog No. 50-002-36415 Supplier Medchemexpress LLC Supplier No. HY1234710MM 1ML
Ro 61-8048 is a selective, orally active inhibitor of kynurenine 3-hydroxylase (KMO) used in biochemical and neurological research to modulate the kynurenine pathway. It has a molecular formula of C17H15N3O6S2 and a molecular weight of approximately 421.5 g/mol, and is supplied as a solid or as a 10 mM solution in DMSO for in vitro use.
  • Selective KMO inhibitor with reported IC50 ≈ 37 nM.
  • High purity suitable for research applications.
  • Available as solid (multiple weights) and as a 10 mM solution in DMSO.
  • Off-white to light yellow solid physical form.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
  • Suitable for studies of the kynurenine pathway and neurological disease models.