Learn More
Medchemexpress LLC 3,4-dimethoxy-N-(4-(3-nitrophenyl)thiazol-2-yl)benzenesulfonamide | 199666-03-0 | MFCD11040807 | 99.8% | 421.45 g/mol | C17H15N3O6S2 | 200MG
SDP
Catalog No. 5000238596
Encompass_Preferred
Ro 61-8048 is a potent, selective kynurenine 3-hydroxylase (kynurenine 3-monooxygenase, KMO) inhibitor used in biochemical and cellular research to modulate the kynurenine pathway. The compound is supplied as a solid research chemical with a reported IC50 ≈ 37 nM, molecular weight ~421.45 g/mol, and high purity suitable for pharmacology studies.
  • Potent inhibitor of KMO with reported IC50 ≈ 37 nM.
  • High purity suitable for biochemical and cellular assays.
  • Solid form with stable storage characteristics for laboratory use.
  • Used to modulate the kynurenine pathway in pharmacology studies.
  • Available in multiple small-mass pack sizes for research applications.
Catalog No. 50-002-38596 Supplier Medchemexpress LLC Supplier No. HY12347200MG
Ro 61-8048 is a potent, selective kynurenine 3-hydroxylase (kynurenine 3-monooxygenase, KMO) inhibitor used in biochemical and cellular research to modulate the kynurenine pathway. The compound is supplied as a solid research chemical with a reported IC50 ≈ 37 nM, molecular weight ~421.45 g/mol, and high purity suitable for pharmacology studies.
  • Potent inhibitor of KMO with reported IC50 ≈ 37 nM.
  • High purity suitable for biochemical and cellular assays.
  • Solid form with stable storage characteristics for laboratory use.
  • Used to modulate the kynurenine pathway in pharmacology studies.
  • Available in multiple small-mass pack sizes for research applications.