Learn More
Medchemexpress LLC 3',4'-dimethoxy-α-naphthoflavone | 14756-24-2 | MFCD00143008 | 98.5% | 332.35 | C21H16O4 | 25 MG
SDP
Catalog No. 5000246026
Encompass_Preferred
DiMNF (3',4'-Dimethoxy-αNF) is a selective aryl hydrocarbon receptor (AHR) modulator used in laboratory research. It acts as a competitive AHR ligand with a reported IC50 = 21 nM and exhibits apparent antagonistic activity; it is applied in studies of AHR-mediated signaling and inflammatory responses. For research use only.
  • Competitive AHR ligand (IC50 = 21 nM).
  • Used to study AHR signaling and inflammation.
  • High purity (98.46%).
  • Molecular formula C21H16O4; molecular weight 332.35.
  • Available as solids (including 25 mg) and as 10 mM solutions in DMSO.
  • Intended for laboratory research only; not for human use.
Catalog No. 50-002-46026 Supplier Medchemexpress LLC Supplier No. HY10322225MG
DiMNF (3',4'-Dimethoxy-αNF) is a selective aryl hydrocarbon receptor (AHR) modulator used in laboratory research. It acts as a competitive AHR ligand with a reported IC50 = 21 nM and exhibits apparent antagonistic activity; it is applied in studies of AHR-mediated signaling and inflammatory responses. For research use only.
  • Competitive AHR ligand (IC50 = 21 nM).
  • Used to study AHR signaling and inflammation.
  • High purity (98.46%).
  • Molecular formula C21H16O4; molecular weight 332.35.
  • Available as solids (including 25 mg) and as 10 mM solutions in DMSO.
  • Intended for laboratory research only; not for human use.