Learn More
Medchemexpress LLC 3',4'-Dimethoxy-α-naphthoflavone | 14756-24-2 | MFCD00143008 | ≥99.0% | 332.35 g·mol⁻1 | C21H16O4 | 100 MG
SDP
Catalog No. 5000247701
Encompass_Preferred
DiMNF (3',4'-Dimethoxy-α-naphthoflavone) is a small-molecule selective aryl hydrocarbon receptor (AHR) modulator used in biochemical and pharmacological research to probe AHR-related signaling. It is supplied as a solid or as a DMSO solution for in vitro assays, with common pack sizes including 100 mg.
  • Selective aryl hydrocarbon receptor modulator for AHR pathway studies.
  • High chemical purity suitable for analytical and biological assays.
  • Molecular formula C21H16O4 and molecular weight 332.35 g·mol⁻1.
  • Available as powder and as 10 mM solution in DMSO for convenient dosing.
  • Stable under recommended storage conditions (2-8 °C) for long-term use.
Catalog No. 50-002-47701 Supplier Medchemexpress LLC Supplier No. HY103222100MG
DiMNF (3',4'-Dimethoxy-α-naphthoflavone) is a small-molecule selective aryl hydrocarbon receptor (AHR) modulator used in biochemical and pharmacological research to probe AHR-related signaling. It is supplied as a solid or as a DMSO solution for in vitro assays, with common pack sizes including 100 mg.
  • Selective aryl hydrocarbon receptor modulator for AHR pathway studies.
  • High chemical purity suitable for analytical and biological assays.
  • Molecular formula C21H16O4 and molecular weight 332.35 g·mol⁻1.
  • Available as powder and as 10 mM solution in DMSO for convenient dosing.
  • Stable under recommended storage conditions (2-8 °C) for long-term use.