Learn More
Medchemexpress LLC 3',4'-Dimethoxy-αNF | 14756-24-2 | MFCD00143008 | 98.5% | 332.35 | C21H16O4 | 5 MG
SDP
Catalog No. 5000263689
Encompass_Preferred
DiMNF (3',4'-Dimethoxy-αNF) is a selective aryl hydrocarbon receptor (AHR) modulator provided as a research reagent. It is supplied as a light yellow solid or as DMSO solutions and intended for laboratory research use only.
  • Selective aryl hydrocarbon receptor modulator used in AHR signaling studies.
  • High purity (≈98.5%) suitable for biochemical assays.
  • Molecular weight 332.35 and formula C21H16O4 for reference.
  • Available in small solid quantities and as 10 mM DMSO solutions.
  • Light yellow solid appearance for easy physical identification.
Catalog No. 50-002-63689 Supplier Medchemexpress LLC Supplier No. HY1032225MG
DiMNF (3',4'-Dimethoxy-αNF) is a selective aryl hydrocarbon receptor (AHR) modulator provided as a research reagent. It is supplied as a light yellow solid or as DMSO solutions and intended for laboratory research use only.
  • Selective aryl hydrocarbon receptor modulator used in AHR signaling studies.
  • High purity (≈98.5%) suitable for biochemical assays.
  • Molecular weight 332.35 and formula C21H16O4 for reference.
  • Available in small solid quantities and as 10 mM DMSO solutions.
  • Light yellow solid appearance for easy physical identification.