Learn More
Medchemexpress LLC 3',4'-dimethoxy-αNF | 14756-24-2 | MFCD00143008 | 98.5% | 332.35 g/mol | C21H16O4 | 1 ML
SDP
Catalog No. 5000247049
Encompass_Preferred
DiMNF (3',4'-Dimethoxy-αNF) is a small-molecule aryl hydrocarbon receptor (AHR) ligand provided for research use. It is supplied as a 10 mM solution in DMSO (1 mL) and is also available as solid quantities for biochemical and cellular assays.
  • Small-molecule AHR modulator for in vitro research.
  • Provided as a 10 mM solution in DMSO suitable for cellular assays.
  • High purity for reliable experimental results.
  • Available in multiple solid sizes for storage or custom use.
  • Molecular weight 332.35 g/mol and formula C21H16O4 for reference.
Catalog No. 50-002-47049 Supplier Medchemexpress LLC Supplier No. HY10322210MM 1ML
DiMNF (3',4'-Dimethoxy-αNF) is a small-molecule aryl hydrocarbon receptor (AHR) ligand provided for research use. It is supplied as a 10 mM solution in DMSO (1 mL) and is also available as solid quantities for biochemical and cellular assays.
  • Small-molecule AHR modulator for in vitro research.
  • Provided as a 10 mM solution in DMSO suitable for cellular assays.
  • High purity for reliable experimental results.
  • Available in multiple solid sizes for storage or custom use.
  • Molecular weight 332.35 g/mol and formula C21H16O4 for reference.