Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxyaniline, Spectrum™ Chemical
SDP

Catalog No. 18620569 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-569 25 g
18-620-570 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-569 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D155825GM
Add to Cart
Add to Cart

Synonym: 4-Aminoveratrole, 4-Aminoveratrol, 3,4-Dimethoxy-Benzenamine

Chemical Identifiers

CAS 6315-89-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
InChI Key LGDHZCLREKIGKJ-UHFFFAOYSA-N
IUPAC Name 3,4-dimethoxyaniline
SMILES COC1=CC=C(N)C=C1OC

Specifications

Boiling Point 180°C
Packaging Amber Bottle
Quantity 25 g
Percent Purity 98%
Physical Form Crystals or Flakes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.