Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-Dimethoxybenzamide | 1521-41-1 | 181.19 | 50 MG
SDP

Catalog No. 5000415234
Encompass_Preferred

3,4-Dimethoxybenzamide is an amide isolated from the solid culture of Streptoverticillium morookaense. It serves as a starting material for the preparation of Itopride hydrochloride.

  • Purity of 98.84%
  • Molecular weight of 181.19
  • Chemical formula: C9H11NO3
  • Appears as a solid, white to off-white in color
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Solubility in DMSO: 125 mg/mL

Catalog No. 50-004-15234 Supplier Medchemexpress LLC Supplier No. HYN177750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,4-Dimethoxybenzamide is an amide isolated from the solid culture of Streptoverticillium morookaense. It serves as a starting material for the preparation of Itopride hydrochloride.

  • Purity of 98.84%
  • Molecular weight of 181.19
  • Chemical formula: C9H11NO3
  • Appears as a solid, white to off-white in color
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Solubility in DMSO: 125 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.