Learn More
Medchemexpress LLC 3,4-Dimethoxycinnamic acid | 2316-26-9 | 208.21 | 1 G
SDP
Catalog No. 5000415193
Encompass_Preferred
3,4-Dimethoxycinnamic acid (O-Methylferulic acid) is a monomer extracted and purified from Securidaca inappendiculata Hassk. It exerts anti-apoptotic effects on L-02 cells via the ROS-mediated signaling pathway.
  • Attenuates the ethanol-induced apoptosis of ethanol-exposed L-02 cells
  • Inhibits the expression levels of Nox4 and p22phox in L-02 cells
  • Attenuates ethanol-induced MAPK phosphorylation in L-02 cells
  • Decreases the expression levels of superoxide dismutase, catalase and phospholipid hydroperoxide gluthione peroxidase
  • Downregulates the levels of Bax/Bcl-2 and the cleaved forms of caspase-3 in a dose- and time-dependent manner
Catalog No. 50-004-15193 Supplier Medchemexpress LLC Supplier No. HYN17781G
3,4-Dimethoxycinnamic acid (O-Methylferulic acid) is a monomer extracted and purified from Securidaca inappendiculata Hassk. It exerts anti-apoptotic effects on L-02 cells via the ROS-mediated signaling pathway.
  • Attenuates the ethanol-induced apoptosis of ethanol-exposed L-02 cells
  • Inhibits the expression levels of Nox4 and p22phox in L-02 cells
  • Attenuates ethanol-induced MAPK phosphorylation in L-02 cells
  • Decreases the expression levels of superoxide dismutase, catalase and phospholipid hydroperoxide gluthione peroxidase
  • Downregulates the levels of Bax/Bcl-2 and the cleaved forms of caspase-3 in a dose- and time-dependent manner