Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxyphenethyl isothiocyanate, 95%, Thermo Scientific™

Catalog No. TL00669DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TL00669DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00669DA Supplier Thermo Scientific Chemicals Supplier No. TL00669DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21714-25-0
Molecular Formula C11H13NO2S
Molecular Weight (g/mol) 223.29
MDL Number MFCD00041366
InChI Key GHYFLDWHIVKQLS-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenethyl isothiocyanate, 4-2-isothiocyanatoethyl-1,2-dimethoxybenzene, 2-3,4-dimethoxyphenyl ethyl isothiocyanate, benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy, 2-3,4-dimethoxyphenyl ethanisothiocyanate, acmc-20anep, 3,4-dimethoxyphenethylisothiocyanate, b-3,4-dimethoxyphenyl ethyl isothiocyanate, 3,4-dimethoxy-1-2-isothiocyanatoethyl benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy-benzene
PubChem CID 140856
IUPAC Name 4-(2-isothiocyanatoethyl)-1,2-dimethoxybenzene
SMILES COC1=CC=C(CCN=C=S)C=C1OC

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 223.29
Chemical Name or Material 3, 4-Dimethoxyphenethyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.