Learn More
Medchemexpress LLC 3,4-Dimethoxyphenol | 2033-89-8 | 154.16 | 50 G
SDP
Catalog No. 5000416322
Encompass_Preferred
3,4-Dimethoxyphenol is a plant-derived phenylpropanoid compound that can be used as a whitening agent in cosmetics. It exhibits broad-spectrum antimicrobial activity, tyrosinase-inhibiting activity, and a potent antioxidant effect, isolated from bacterial fermentation broth. It can be used for the study of infection.
  • Induces morphological damage in Xoo cells (62.5-125 μg/mL in vitro).
Catalog No. 50-004-16322 Supplier Medchemexpress LLC Supplier No. HYN178050G
3,4-Dimethoxyphenol is a plant-derived phenylpropanoid compound that can be used as a whitening agent in cosmetics. It exhibits broad-spectrum antimicrobial activity, tyrosinase-inhibiting activity, and a potent antioxidant effect, isolated from bacterial fermentation broth. It can be used for the study of infection.
  • Induces morphological damage in Xoo cells (62.5-125 μg/mL in vitro).