Learn More
Medchemexpress LLC 3,4-Dimethoxyphenol | 2033-89-8 | MFCD00008390 | 154.16 | 10 G
SDP
Catalog No. 5000415394
Encompass_Preferred
3,4-Dimethoxyphenol is a plant-derived phenylpropanoid compound that can be used as a whitening agent in cosmetics. It demonstrates broad-spectrum antimicrobial activity, tyrosinase-inhibiting activity, and potent antioxidant effects. It is isolated from bacterial fermentation broth and can be utilized for the study of infection.
  • Plant-derived phenylpropanoid compound
  • Whitening agent in cosmetics
  • Broad-spectrum antimicrobial activity
  • Tyrosinase-inhibiting activity
  • Potent antioxidant effect
  • Can be used for the study of infection
Catalog No. 50-004-15394 Supplier Medchemexpress LLC Supplier No. HYN178010G
3,4-Dimethoxyphenol is a plant-derived phenylpropanoid compound that can be used as a whitening agent in cosmetics. It demonstrates broad-spectrum antimicrobial activity, tyrosinase-inhibiting activity, and potent antioxidant effects. It is isolated from bacterial fermentation broth and can be utilized for the study of infection.
  • Plant-derived phenylpropanoid compound
  • Whitening agent in cosmetics
  • Broad-spectrum antimicrobial activity
  • Tyrosinase-inhibiting activity
  • Potent antioxidant effect
  • Can be used for the study of infection