Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3,4-Dimethoxyphenyl)(4-morpholinyl)acetonitrile

Catalog No. OTV000031 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000031 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000031 Supplier Sigma Aldrich Supplier No. OTV000031250MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-(trimethylsilyl)-3-butyn-2-one is a ketone
  • Its asymmetric bioreduction to enantiopure {(s)-tmsbol in various hydrophilic ionic liquid (ils) solvent systems has been reported.4-(trimethylsilyl)-3-butyn-2-one (tmsb) has been used to investigate its asymmetric bioreduction to (s)-4-(trimethylsilyl)-3-butyn-2-ol {(s)-tmsbol} by employing biocompatible water-immiscible ionic liquids (ils)
  • Tmsb may be used for the synthesis of entecavir (bms-200475)

Specifications

Molecular Formula C14H18N2O3
Linear Formula C14H18N2O3
MDL Number MFCD09702581
Quantity 5 g
Molecular Weight (g/mol) 262.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.