Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (3,4-Dimethoxyphenyl)hydrazine hydrochloride | 40119-17-3 | 204.66 | 100 MG
SDP

Catalog No. 5000447318
Encompass_Preferred

(3,4-Dimethoxyphenyl)hydrazine hydrochloride is a drug intermediate used for the synthesis of various active compounds.

  • Used for the synthesis of various active compounds
  • Purity 99.84%
  • Molecular weight 204.66
  • Appearance: solid
  • Color: light brown to gray

Catalog No. 50-004-47318 Supplier Medchemexpress LLC Supplier No. HYW006381100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(3,4-Dimethoxyphenyl)hydrazine hydrochloride is a drug intermediate used for the synthesis of various active compounds.

  • Used for the synthesis of various active compounds
  • Purity 99.84%
  • Molecular weight 204.66
  • Appearance: solid
  • Color: light brown to gray

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.