Learn More
Medchemexpress LLC (3,4-Dimethoxyphenyl)hydrazine (hydrochloride) | 40119-17-3 | 204.66 | 250 MG
SDP
Catalog No. 5000450036
Encompass_Preferred
(3,4-Dimethoxyphenyl)hydrazine hydrochloride is a chemical compound primarily utilized as a drug intermediate for the synthesis of various active compounds. This product is intended for research use only and is not for human use.
  • Used as a drug intermediate
  • Aids in the synthesis of active compounds
  • High purity of 99.8%
  • Solid appearance
  • Light brown to gray color
  • For research use only
Catalog No. 50-004-50036 Supplier Medchemexpress LLC Supplier No. 250MGHYW006381
(3,4-Dimethoxyphenyl)hydrazine hydrochloride is a chemical compound primarily utilized as a drug intermediate for the synthesis of various active compounds. This product is intended for research use only and is not for human use.
  • Used as a drug intermediate
  • Aids in the synthesis of active compounds
  • High purity of 99.8%
  • Solid appearance
  • Light brown to gray color
  • For research use only