Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4-Dimethoxythiophene

Catalog No. 6682575G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-825-75G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66-825-75G Supplier Sigma Aldrich Supplier No. 6682575G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,4-Dimethoxythiophene (DMOT) is a monomer and a precursor which can be synthesized by ring closure reaction of 2,3-dimethoxy-1,3-butadiene and sulfur dichloride in hexane medium. It is an oligothiphene that is majorly used in the development of electroactive materials for organic electronics based applications.DMOT can be trans-esterified to form 3,4-ethylenendioxythiophene (EDOT). It can further be polymerized to produce PEDOT which can be used as a conductive polymer inconjugated systems. It can be polymerized to form poly(dimethoxythiphenes) which can potentially be used in the fabrication energy storage devices on electrochemical doping.

Specifications

CAS 51792-34-8
Density 1.209 g/mL (at 25°C)
Boiling Point 100°C to 102°C/10°C to 11 mmHg
Molecular Formula C6H8O2S
Linear Formula C6H8O2S
Refractive Index n20/D 1.5409
MDL Number MFCD01096546
Quantity 5 g
Molecular Weight (g/mol) 144.19
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.