Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxythiophenol, 99+%, Thermo Scientific™

Catalog No. AC268060100
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC268060100 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC268060100 Supplier Thermo Scientific Chemicals Supplier No. 268060100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 700-96-9
CAS Min % 99.0
CAS Max % 100.0
Density 1.1900g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C8H10O2S
Refractive Index 1.5890 to 1.5910 (20°C, 589nm)
Quantity 10g
Synonym 3,4-dimethoxythiophenol, 3,4-dimethoxy thiophenol, 3,4-dimethoxybenzene-1-thiol, 3,4-dimethoxy-thiophenol, benzenethiol, 3,4-dimethoxy, 4-mercaptoveratrole, pubchem6823, acmc-1bdpf, 3,4-dimethoxy-benzenethiol, 3,4-dimethoxyphenylmercaptan
InChI Key MTKAJLNGIVXZIS-UHFFFAOYSA-N
SMILES COC1=C(C=C(C=C1)S)OC
IUPAC Name 3,4-dimethoxybenzenethiol
Molecular Weight (g/mol) 170.226
PubChem CID 136539
Formula Weight 170.23
Percent Purity ≥99.0%
Chemical Name or Material 3, 4-Dimethoxythiophenol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.