Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethyl-3-cyclohexenecarboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. D5098200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5098200MG 200 mg
D50981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5098200MG Supplier TCI America Supplier No. D5098200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18022-66-7
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00060823
InChI Key NOJNKSAQZMVMFF-UHFFFAOYSA-N
PubChem CID 537551
IUPAC Name 3,4-dimethylcyclohex-3-ene-1-carbaldehyde
SMILES CC1=C(CC(CC1)C=O)C

Specifications

Color Colorless
Boiling Point 79°C
Quantity 200 mg
Formula Weight 138.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethyl-3-cyclohexenecarboxaldehyde
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.