Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethyl-3-hexanol 99.0+%, TCI America™
SDP

Catalog No. D197010ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D197010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D197010ML Supplier TCI America Supplier No. D197010ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19550-08-4
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00021820
InChI Key FJXOYCIYKQJAAF-UHFFFAOYSA-N
PubChem CID 140548
IUPAC Name 3,4-dimethylhexan-3-ol
SMILES CCC(C)C(C)(CC)O

Specifications

Color Colorless
Boiling Point 58°C
Quantity 10 mL
UN Number 1987
Formula Weight 130.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethyl-3-hexanol
EINECSNumber (2)-0217
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.