Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',4'-Dimethylacetanilide 98.0+%, TCI America™
SDP

Catalog No. D212210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D212210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D212210G Supplier TCI America Supplier No. D212210G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2198-54-1
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00026137
InChI Key UAOIEEWQVAXCFY-UHFFFAOYSA-N
Synonym 3',4'-dimethylacetanilide, n-3,4-dimethylphenyl acetamide, 3,4-dimethylacetanilide, n-acetyl-3,4-xylidine, 3',4'-acetoxylidide, acetamide, n-3,4-dimethylphenyl, 3',4'-acetoxylide, acetamide, n-3,4-dimethylphenyl-9ci, 4-acetamino-o-xylol, aceto-3,4-xylidide
PubChem CID 75144
IUPAC Name N-(3,4-dimethylphenyl)acetamide
SMILES CC1=C(C=C(C=C1)NC(=O)C)C

Specifications

Melting Point 98°C
Quantity 10 g
Formula Weight 163.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,4′-Dimethylacetanilide
EINECSNumber (3)-0269
RTECSNumber AN7500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.