Learn More
Medchemexpress LLC 3-[4-[(Dimethylamino)methyl]phenyl]-N-methyl-N-[5-[3-methyl-5-(1,3,5-trimethyl-1H-pyrazol-4-yl)-2-py | 2830555-70-7 | >97.0% | 561.68 g·mol⁻¹ | C32H35N9O | 5 MG
SDP
Catalog No. 5000262915
Encompass_Preferred
A selective type I1/2 dual Mer and Axl receptor tyrosine kinase inhibitor for preclinical biochemical, cellular, and in vivo research. Molecular formula C32H35N9O with a molecular weight of 561.68 g·mol⁻¹. Supplied as small-mass solids and DMSO solutions with documented solubility and in vivo vehicle options.
  • Highly selective type I1/2 dual Mer/Axl kinase inhibitor.
  • Demonstrated in vivo efficacy in preclinical models.
  • Supplied as solid and pre-made DMSO solution formats.
  • Soluble at ≥ 5 mg/mL in recommended vehicles.
  • High purity (≥97% by HPLC).
  • Suitable for biochemical, cellular, and pharmacology studies.
Catalog No. 50-002-62915 Supplier Medchemexpress LLC Supplier No. HY1328935MG
A selective type I1/2 dual Mer and Axl receptor tyrosine kinase inhibitor for preclinical biochemical, cellular, and in vivo research. Molecular formula C32H35N9O with a molecular weight of 561.68 g·mol⁻¹. Supplied as small-mass solids and DMSO solutions with documented solubility and in vivo vehicle options.
  • Highly selective type I1/2 dual Mer/Axl kinase inhibitor.
  • Demonstrated in vivo efficacy in preclinical models.
  • Supplied as solid and pre-made DMSO solution formats.
  • Soluble at ≥ 5 mg/mL in recommended vehicles.
  • High purity (≥97% by HPLC).
  • Suitable for biochemical, cellular, and pharmacology studies.