Learn More
Medchemexpress LLC 3-[4-[(dimethylamino)methyl]phenyl]-N-methyl-N-[5-[3-methyl-5-(1,3,5-trimethyl-1H-pyrazol-4-yl)-2-pyri... | 2830555-70-7 | 98.0% | C32H35N9O | 10MG
SDP
Catalog No. 5000259778
Encompass_Preferred
AZ14145845 is a selective type I1/2 dual Mer and Axl kinase inhibitor developed for research use with demonstrated in vivo efficacy. It is supplied as a white to off-white solid, has molecular formula C32H35N9O and molecular weight 561.68, and is provided at high purity for preclinical and laboratory applications.
  • Selective inhibition of Mer and Axl kinases.
  • Demonstrated in vivo efficacy in preclinical models.
  • High chemical purity suitable for biochemical and cellular assays.
  • Available in small-scale quantities and solution formulations for screening.
  • White to off-white solid with standard laboratory storage characteristics.
Catalog No. 50-002-59778 Supplier Medchemexpress LLC Supplier No. HY13289310MG
AZ14145845 is a selective type I1/2 dual Mer and Axl kinase inhibitor developed for research use with demonstrated in vivo efficacy. It is supplied as a white to off-white solid, has molecular formula C32H35N9O and molecular weight 561.68, and is provided at high purity for preclinical and laboratory applications.
  • Selective inhibition of Mer and Axl kinases.
  • Demonstrated in vivo efficacy in preclinical models.
  • High chemical purity suitable for biochemical and cellular assays.
  • Available in small-scale quantities and solution formulations for screening.
  • White to off-white solid with standard laboratory storage characteristics.