Learn More
Medchemexpress LLC 3-(4-(dimethylamino)phenyl)acrylaldehyde | 6203-18-5 | MFCD00007002 | 99.8% | 175.23 | C11H13NO | 100 G
SDP
Catalog No. 5000303240
Encompass_Preferred
4-(Dimethylamino)cinnamaldehyde is an aromatic aldehyde reagent used in biochemical assays to detect indole-containing compounds and to quantify proanthocyanidins in plant extracts. Supplied as a powder with high analytical purity, it is suitable for research and assay applications.
  • Reagent for indole detection and proanthocyanidin quantification.
  • High purity (99.8%) suitable for analytical use.
  • Powder form for convenient storage and handling.
  • Stable under recommended storage: -20°C for powder or low-temperature solvent storage.
  • Molecular formula C11H13NO; molecular weight 175.23 g/mol.
Catalog No. 50-003-03240 Supplier Medchemexpress LLC Supplier No. HYW016729100G
4-(Dimethylamino)cinnamaldehyde is an aromatic aldehyde reagent used in biochemical assays to detect indole-containing compounds and to quantify proanthocyanidins in plant extracts. Supplied as a powder with high analytical purity, it is suitable for research and assay applications.
  • Reagent for indole detection and proanthocyanidin quantification.
  • High purity (99.8%) suitable for analytical use.
  • Powder form for convenient storage and handling.
  • Stable under recommended storage: -20°C for powder or low-temperature solvent storage.
  • Molecular formula C11H13NO; molecular weight 175.23 g/mol.