Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4-Dimethylbenzaldehyde

Catalog No. 49385625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-385-625G 25 g
49-385-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 49-385-625G Supplier Sigma Aldrich Supplier No. 49385625G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,4-dimethylbenzaldehyde (3,4-dmb) is a benzaldehyde derivative
  • It is the oh radical initiated oxidative degradation product of trimethylbenzene
  • The rate coefficient of the gas-phase reaction between 3,4-dmb and oh radical is 24.6+/-4.0x10-12cm3molecule-1s-1
  • The vibrational analysis of 3,4-dmb based on ft-IR spectra, ft-raman spectra, ab initio and density functional theory (dft) calculations have been reported
  • It is formed as an intermediate during the transformation of furfural into gasoline-range fuels using zsm(zeolite socony mobil)-5-based catalysts.3,4-dimethylbenzaldehyde may be used in the synthesis of 3,4-dimethylmethcathinone (dmmc) and 3,4-dimethyl-dibenzylidene sorbitol

Specifications

CAS 5973-71-7
Density 1.012 g/mL (at 25°C (literature))
Boiling Point 226°C (lit.)
Molecular Formula C9H10O
Refractive Index n20/D 1.551 (literature)
Linear Formula (CH3)2C6H3CHO
MDL Number MFCD00016612
Quantity 25 g
Molecular Weight (g/mol) 134.18
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.