Learn More
Medchemexpress LLC 3,4-dimethylbenzoic acid | 619-04-5 | MFCD00002524 | 99.8% | 150.18 g/mol | C9H10O2 | 1 ML
SDP
Catalog No. 5000300182
Encompass_Preferred
3,4-Dimethylbenzoic acid is a benzoic acid derivative used as a research standard and metabolite reference. It is provided as a solid or as a ready-to-use 10 mM solution in DMSO for analytical and metabolic studies, and is accompanied by batch documentation to support experimental reproducibility.
  • High purity suitable for analytical applications (reported ~99.8%).
  • Available as a 10 mM solution in DMSO and as solid material in multiple pack sizes.
  • Well-characterized molecular properties (C9H10O2; ~150.18 g/mol).
  • Suitable as a metabolite reference in biodegradation and metabolic pathway studies.
  • Batch-specific certificate of analysis available for quality assurance.
Catalog No. 50-003-00182 Supplier Medchemexpress LLC Supplier No. HYW01743410MM/1ML
3,4-Dimethylbenzoic acid is a benzoic acid derivative used as a research standard and metabolite reference. It is provided as a solid or as a ready-to-use 10 mM solution in DMSO for analytical and metabolic studies, and is accompanied by batch documentation to support experimental reproducibility.
  • High purity suitable for analytical applications (reported ~99.8%).
  • Available as a 10 mM solution in DMSO and as solid material in multiple pack sizes.
  • Well-characterized molecular properties (C9H10O2; ~150.18 g/mol).
  • Suitable as a metabolite reference in biodegradation and metabolic pathway studies.
  • Batch-specific certificate of analysis available for quality assurance.