Learn More
Medchemexpress LLC 3,4-dimethylbenzoic acid | 619-04-5 | MFCD00002524 | 99.8% | 150.18 g/mol | C9H10O2 | 100 G
SDP
Catalog No. 5000301379
Encompass_Preferred
3,4-Dimethylbenzoic acid (CAS 619-04-5) is a benzoic acid derivative and reported product of dimethylbenzoate metabolism. Supplied as a white-to-yellow solid powder, it is used as a metabolite reference standard and analytical reagent for research. Molecular formula C9H10O2; molecular weight 150.18 g/mol.
  • High purity (99.79%) suitable for analytical and metabolite studies.
  • Solid powder form for ease of weighing and storage.
  • Soluble in DMSO at 100 mg/mL (≈665.87 mM) for in vitro preparations.
  • Long-term stability when stored as powder (-20°C for up to 3 years, 4°C for up to 2 years).
  • Appropriate as a reference standard in metabolite identification and analytical chemistry.
Catalog No. 50-003-01379 Supplier Medchemexpress LLC Supplier No. HYW017434100G
3,4-Dimethylbenzoic acid (CAS 619-04-5) is a benzoic acid derivative and reported product of dimethylbenzoate metabolism. Supplied as a white-to-yellow solid powder, it is used as a metabolite reference standard and analytical reagent for research. Molecular formula C9H10O2; molecular weight 150.18 g/mol.
  • High purity (99.79%) suitable for analytical and metabolite studies.
  • Solid powder form for ease of weighing and storage.
  • Soluble in DMSO at 100 mg/mL (≈665.87 mM) for in vitro preparations.
  • Long-term stability when stored as powder (-20°C for up to 3 years, 4°C for up to 2 years).
  • Appropriate as a reference standard in metabolite identification and analytical chemistry.