Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4-Dimethylbenzylamine

Catalog No. CDS002514 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS002514 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS002514 Supplier Sigma Aldrich Supplier No. CDS00251450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-bromoheptadecafluorooctane (perfluorooctyl bromide) is a perfluorocarbon
  • Poly(ethylene glycol)ylated polyester nanocapsules of perfluorooctyl bromide were surface-decorated with a rgd (arginine-glycine-aspartic acid) peptide, were used for tumor imaging
  • Nanocapsules of perfluorooctyl bromide have been developed as dual contrast agent for ultrasonography and magnetic resonance imaging (mri).1-bromoheptadecafluorooctane (perfluorooctyl bromide) can be used as contrast agent in the vascular system for fluorine-19 magnetic resonance imaging or as synthetic oxygen carrier
  • It was also employed as oxygen carrier in the emulsions used as blood substitutes

Specifications

CAS 102-48-7
Molecular Formula C9H13N
Linear Formula C9H13N
MDL Number MFCD00025577
Quantity 5 g
Molecular Weight (g/mol) 135.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.