Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethylphenylhydrazine hydrochloride, 98%

Catalog No. AAA1302730
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1302730 250 g
AAA1302709 10 g
AAA1302718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1302730 Supplier Thermo Scientific Chemicals Supplier No. A1302730
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 60481-51-8 | C8H13ClN2 | 172.656 g/mol

3,4-Dimethylphenylhydrazine hydrochloride, is used as a reactant with ethyl 3-ethoxy-2-ethoxycarbonylprop-2-enoate to produce 2-(3,4-dimethyl-phenyl)-3-oxo-2,3-dihydro-1H-pyrazole-4-carboxylic acid ethyl ester. This reaction will need reagent K2CO3, and the menstruum ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylphenylhydrazine hydrochloride, is used as a reactant with ethyl 3-ethoxy-2-ethoxycarbonylprop-2-enoate to produce 2-(3,4-dimethyl-phenyl)-3-oxo-2,3-dihydro-1H-pyrazole-4-carboxylic acid ethyl ester. This reaction will need reagent K2CO3, and the menstruum ethanol.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60481-51-8
Molecular Formula C8H13ClN2
Molecular Weight (g/mol) 172.656
MDL Number MFCD00052270
InChI Key YYMIOVAEQIEPET-UHFFFAOYSA-N
Synonym 3,4-dimethylphenylhydrazine hydrochloride, 3,4-dimethylphenyl hydrazine hydrochloride, 1-3,4-dimethylphenyl hydrazine hydrochloride, hydrazine, 3,4-dimethylphenyl-, monohydrochloride, 4-hydrazino-o-xylene hydrochloride, 3,4-dimethylphenyl hydrazine hydrochloride 1:x, hydrazine, 3,4-dimethylphenyl-, hydrochloride, 3,4-dimethylphenyl diazane hydrochloride, pubchem7555, 3,4-dimethylphenylhydrazinehydrochloride
PubChem CID 173740
IUPAC Name (3,4-dimethylphenyl)hydrazine;hydrochloride
SMILES CC1=C(C=C(C=C1)NN)C.Cl

Specifications

Melting Point ∼195°C (decomposition)
Quantity 250 g
Beilstein 6096060
Solubility Information Soluble in water.
Formula Weight 172.66
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylphenylhydrazine hydrochloride
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.