Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethylphenylhydrazine Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. D36165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36165G 5 g
D361625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36165G Supplier TCI America Supplier No. D36165G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60481-51-8
Molecular Formula C8H13ClN2
Molecular Weight (g/mol) 172.656
MDL Number MFCD02656652
InChI Key YYMIOVAEQIEPET-UHFFFAOYSA-N
Synonym 3,4-dimethylphenylhydrazine hydrochloride, 3,4-dimethylphenyl hydrazine hydrochloride, 1-3,4-dimethylphenyl hydrazine hydrochloride, hydrazine, 3,4-dimethylphenyl-, monohydrochloride, 4-hydrazino-o-xylene hydrochloride, 3,4-dimethylphenyl hydrazine hydrochloride 1:x, hydrazine, 3,4-dimethylphenyl-, hydrochloride, 3,4-dimethylphenyl diazane hydrochloride, pubchem7555, 3,4-dimethylphenylhydrazinehydrochloride
PubChem CID 173740
IUPAC Name (3,4-dimethylphenyl)hydrazine;hydrochloride
SMILES CC1=C(C=C(C=C1)NN)C.Cl

Specifications

Melting Point 202°C
Quantity 5 g
Formula Weight 172.66
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethylphenylhydrazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.