Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-dimethylpyrazole phosphate | 202842-98-6 | MFCD09842544 | 98.0% | 194.13 g/mol | C5H11N2O4P | 1 KG
SDP

Catalog No. 5000299947
Encompass_Preferred

3,4-dimethylpyrazole phosphate is the phosphate salt of 3,4-dimethyl-1H-pyrazole used as a nitrification inhibitor in research and agricultural studies. It reduces soil nitrification rates by inhibiting ammonia-oxidizing microorganisms and is supplied as a stable, crystalline phosphate salt suitable for formulation development and laboratory testing.

  • Reduces soil nitrification rates in laboratory and formulation studies.
  • Purity 98.0% (NMR), suitable for analytical applications.
  • Molecular weight 194.13 g/mol and formula C5H11N2O4P assist formulation calculations.
  • Stable phosphate salt form for easier handling and storage.
  • Available in multiple package sizes, including 1 kg for scale-up studies.

Catalog No. 50-002-99947 Supplier Medchemexpress LLC Supplier No. HYW0193341KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3,4-dimethylpyrazole phosphate is the phosphate salt of 3,4-dimethyl-1H-pyrazole used as a nitrification inhibitor in research and agricultural studies. It reduces soil nitrification rates by inhibiting ammonia-oxidizing microorganisms and is supplied as a stable, crystalline phosphate salt suitable for formulation development and laboratory testing.

  • Reduces soil nitrification rates in laboratory and formulation studies.
  • Purity 98.0% (NMR), suitable for analytical applications.
  • Molecular weight 194.13 g/mol and formula C5H11N2O4P assist formulation calculations.
  • Stable phosphate salt form for easier handling and storage.
  • Available in multiple package sizes, including 1 kg for scale-up studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.