Learn More
Medchemexpress LLC 3,4-dimethylpyrazole phosphate | 202842-98-6 | MFCD09842544 | 98.0% | 194.13 g/mol | C5H11N2O4P | 1 ML
SDP
Catalog No. 5000299946
Encompass_Preferred
3,4-dimethylpyrazole phosphate (3,4-DMPP) is a nitrification inhibitor used in agricultural and laboratory research. It is provided as a ready-to-use 10 mM solution (1 mL in DMSO) and in bulk solid forms, with a reported purity of 98.0%. The compound is commonly used in studies of nitrification inhibition, fertilizer efficiency, and as an insecticide synergist.
  • Ready-to-use 10 mM solution in DMSO (1 mL)
  • Also available as solid in multiple pack sizes
  • High purity suitable for research applications (98.0%)
  • Used to inhibit nitrification and improve nitrogen use efficiency
  • Compatible with standard analytical and formulation workflows
Catalog No. 50-002-99946 Supplier Medchemexpress LLC Supplier No. HYW01933410MM/1ML
3,4-dimethylpyrazole phosphate (3,4-DMPP) is a nitrification inhibitor used in agricultural and laboratory research. It is provided as a ready-to-use 10 mM solution (1 mL in DMSO) and in bulk solid forms, with a reported purity of 98.0%. The compound is commonly used in studies of nitrification inhibition, fertilizer efficiency, and as an insecticide synergist.
  • Ready-to-use 10 mM solution in DMSO (1 mL)
  • Also available as solid in multiple pack sizes
  • High purity suitable for research applications (98.0%)
  • Used to inhibit nitrification and improve nitrogen use efficiency
  • Compatible with standard analytical and formulation workflows