Learn More
Medchemexpress LLC 3,4-dimethylpyrazole phosphate | 202842-98-6 | MFCD09842544 | 98.0% | 194.13 g/mol | C5H11N2O4P | 500 G
SDP
Catalog No. 5000300092
Encompass_Preferred
3,4-dimethylpyrazole phosphate (3,4-DMPP) is a nitrification inhibitor used in agricultural and research settings to slow the microbial conversion of ammonium to nitrate, improving nitrogen retention in soil and reducing environmental losses. It is supplied as a solid or in solution for incorporation into fertilizer formulations or soil treatments.
Catalog No. 50-003-00092 Supplier Medchemexpress LLC Supplier No. HYW019334500G
3,4-dimethylpyrazole phosphate (3,4-DMPP) is a nitrification inhibitor used in agricultural and research settings to slow the microbial conversion of ammonium to nitrate, improving nitrogen retention in soil and reducing environmental losses. It is supplied as a solid or in solution for incorporation into fertilizer formulations or soil treatments.