Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethylthiophene, 97%

Catalog No. AC450950050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450950050 5 g
AC450950010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450950050 Supplier Thermo Scientific Chemicals Supplier No. 450950050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 632-15-5 | C6H8S | 112.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-15-5
Molecular Formula C6H8S
Molecular Weight (g/mol) 112.19
MDL Number MFCD01764731
InChI Key GPSFYJDZKSRMKZ-UHFFFAOYSA-N
PubChem CID 79089
IUPAC Name 3,4-dimethylthiophene
SMILES CC1=CSC=C1C

Specifications

Boiling Point 144.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Synonym thiophene, 3,4-dimethyl, 3,4-dimethyl-thiophene, 3,4-dimethyl thiophene, thiophene,3,4-dimethyl
Formula Weight 112.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3,4-Dimethylthiophene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H StatementFlammable liquid and vapour.Harmful if swallowed.Harmful in contact with skin.Harmful if inhaled.
Precautionary Statement GHS P StatementKeep away from heat/sparks/open flames/hot surfaces.- No smoking.Wear protective gloves/protective clothing.IF ON SKIN (or hair): Take off immediately all contaminated clothing.Rinse skin with water/shower.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.