Learn More
Cayman Chemical 3,4-DMMA (hydrochloride), Purity: ≥97%, CAS: 70932-18-2, 50 mg
SDP
Catalog No. 502951192
Encompass_Preferred
An MDMA analog that appears to act as a serotonin-norepinephrine-dopamine releasing agent binding to and inhibiting uptake at noradrenalin and serotonin transporters (Kis = 22.8 and 7.7 μM and IC50s = 253.4 and 108 μM, respectively); significantly less potent than MDMA; intended for forensic and research applications
Catalog No. 50-295-1192 Supplier Cayman Chemical Supplier No. 1112850 MG
An MDMA analog that appears to act as a serotonin-norepinephrine-dopamine releasing agent binding to and inhibiting uptake at noradrenalin and serotonin transporters (Kis = 22.8 and 7.7 μM and IC50s = 253.4 and 108 μM, respectively); significantly less potent than MDMA; intended for forensic and research applications