Learn More
MSC Proto Socket Adapter, 1/2 to 3/4 in Drive, 1 ea
Catalog No. 501920297
Encompass
This socket adapter is designed for high-speed applications with pneumatic and electric impact wrenches, allowing for connection between tools with different drive sizes.
  • Suitable for use with pneumatic and electric impact wrenches
  • Pin and o-ring or RET RING® (sold separately) secure tools onto impact wrench for safety and convenience
  • Specially heat treated for impact use to provide long life, safety, and durability
  • Drive adapter type
  • Features a 1/2 in male drive
  • Features a 3/4 in female drive
  • Measures 1-7/8 in overall length
Catalog No. 50-192-0297 Supplier MSC Supplier No. 38234175
This socket adapter is designed for high-speed applications with pneumatic and electric impact wrenches, allowing for connection between tools with different drive sizes.
  • Suitable for use with pneumatic and electric impact wrenches
  • Pin and o-ring or RET RING® (sold separately) secure tools onto impact wrench for safety and convenience
  • Specially heat treated for impact use to provide long life, safety, and durability
  • Drive adapter type
  • Features a 1/2 in male drive
  • Features a 3/4 in female drive
  • Measures 1-7/8 in overall length