Learn More
Cayman Chemical 3 4-EDMAhydrochlorIde 10mg
SDP
Catalog No. 502968576
Encompass_Preferred
An analog of 3,4-MDMA that stimulates the release of serotonin and dopamine from rat brain synaptosomes at 1 µM
Catalog No. 50-296-8576 Supplier Cayman Chemical Supplier No. 1516410 MG
An analog of 3,4-MDMA that stimulates the release of serotonin and dopamine from rat brain synaptosomes at 1 µM