Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4-Ethylenedithiothiophene

Catalog No. 7290781G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-907-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-907-81G Supplier Sigma Aldrich Supplier No. 7290781G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,4-Ethylenedithiothiophene (EDTT) is a dithiin based conducting polymer that is a sulfur analog of 3,4-ethylendioxythiophene (EDOT) that is majorly used in optoelectronic and electrochemical devices.EDTT can be electrochemically oxidized in acetonitrile to form poly(EDTT) films on the surface of the electrodes, which can be used in the fabrication of organic light emitting diodes (OLEDs) and organic photovoltaics (OPVs).

Specifications

CAS 158962-92-6
Density 1.374 g/mL (at 25°C)
Molecular Formula C6H6S3
Refractive Index n20/D 1.685
Linear Formula C6H6S3
Quantity 1 g
MDL Number MFCD23098725
Synonym 3,4-Ethylenedithiothiophene; 2,3-Dihydrothieno[3,4-b][1,4 ]dithiine; 3,4-Ethylenedisulfanylthiophene; EDTT
Molecular Weight (g/mol) 174.31
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.