Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(4-fluorophenyl)propanoic acid | 459-31-4 | MFCD00060327 | 99.7% | 168.16 g/mol | C9H9FO2 | 500 G
SDP

Catalog No. 5000300236
Encompass_Preferred

3-(4-Fluorophenyl)propanoic acid is an organic biochemical reagent and synthetic building block commonly used in medicinal chemistry and life-science research. It is supplied as a white to off-white powder and is suitable for use in organic synthesis, assay development, and intermediate preparation.

  • High purity (99.7%) suitable for analytical and synthetic applications.
  • Molecular weight 168.16 g/mol.
  • Chemical formula C9H9FO2.
  • Solid form with recommended storage at -20°C for long-term stability.
  • Useful as a synthetic intermediate in medicinal chemistry and material science.

Catalog No. 50-003-00236 Supplier Medchemexpress LLC Supplier No. HYW028185500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(4-Fluorophenyl)propanoic acid is an organic biochemical reagent and synthetic building block commonly used in medicinal chemistry and life-science research. It is supplied as a white to off-white powder and is suitable for use in organic synthesis, assay development, and intermediate preparation.

  • High purity (99.7%) suitable for analytical and synthetic applications.
  • Molecular weight 168.16 g/mol.
  • Chemical formula C9H9FO2.
  • Solid form with recommended storage at -20°C for long-term stability.
  • Useful as a synthetic intermediate in medicinal chemistry and material science.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.