Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(4-fluorophenyl)propanoic acid | 459-31-4 | MFCD00060327 | 99.9% | 168.16 g·mol⁻1 | C9H9FO2 | 25 G
SDP

Catalog No. 5000303039
Encompass_Preferred

3-(4-Fluorophenyl)propanoic acid (CAS 459-31-4) is an aromatic carboxylic acid used as a biochemical reagent and a synthetic building block in organic and medicinal chemistry. Supplied as a white to off-white solid with high chemical purity, it is suitable for use in synthetic transformations, assay development, and as a reference material when reproducible, high-quality starting material is required.

  • High purity: 99.88%.
  • Molecular formula: C9H9FO2.
  • Molecular weight: 168.16 g·mol⁻1.
  • White to off-white solid for convenient handling.
  • Available in multiple package sizes, including 25 g.
  • Storage recommendations: powder -20°C (3 years) or 4°C (2 years); in solvent, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-03039 Supplier Medchemexpress LLC Supplier No. 25GHYW028185
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(4-Fluorophenyl)propanoic acid (CAS 459-31-4) is an aromatic carboxylic acid used as a biochemical reagent and a synthetic building block in organic and medicinal chemistry. Supplied as a white to off-white solid with high chemical purity, it is suitable for use in synthetic transformations, assay development, and as a reference material when reproducible, high-quality starting material is required.

  • High purity: 99.88%.
  • Molecular formula: C9H9FO2.
  • Molecular weight: 168.16 g·mol⁻1.
  • White to off-white solid for convenient handling.
  • Available in multiple package sizes, including 25 g.
  • Storage recommendations: powder -20°C (3 years) or 4°C (2 years); in solvent, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.