Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Hexanedione, 94%

Catalog No. AAL0665522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0665522 100 g
AAL0665514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0665522 Supplier Thermo Scientific Chemicals Supplier No. L0665522
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4437-51-8 | C6H10O2 | 114.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4437-51-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00010237
InChI Key KVFQMAZOBTXCAZ-UHFFFAOYSA-N
Synonym 3,4-hexanedione, bipropionyl, dipropionyl, diethyl diketone, 3,4-hexane dione, unii-4m4npr5vgd, 3,4-hexandione, fema no. 3168, ccris 6295, 4m4npr5vgd
PubChem CID 62539
IUPAC Name hexane-3,4-dione
SMILES CCC(=O)C(=O)CC

Specifications

Melting Point -10°C
Density 0.941
Boiling Point 129°C to 131°C
Flash Point 29°C (84°F)
Refractive Index 1.412
Quantity 100 g
UN Number UN1224
Beilstein 1700837
Formula Weight 114.15
Percent Purity 95%
Chemical Name or Material 3,4-Hexanedione
Show More Show Less
Hazard Category H226-H315-H319-H331-H332
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P311-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 224-651-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.