Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(4-Hydroxyphenyl)-1-propanol | 10210-17-0 | 152.19 | 1 ML
SDP

Catalog No. 5000415012
Encompass_Preferred

3-(4-Hydroxyphenyl)-1-propanol (Dihydro-p-coumaryl alcohol) is a metabolite that can be isolated from P. radiate callus cultures and is associated with CAD activity. It is intended for research use only and not for medical applications.

  • Purity of 98.70%
  • Appearance as an off-white to light yellow solid
  • Stable under recommended storage conditions
  • Soluble in DMSO at 25 mg/mL
  • Store powder at -20°C for 3 years, or 4°C for 2 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month

Catalog No. 50-004-15012 Supplier Medchemexpress LLC Supplier No. HYN175310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(4-Hydroxyphenyl)-1-propanol (Dihydro-p-coumaryl alcohol) is a metabolite that can be isolated from P. radiate callus cultures and is associated with CAD activity. It is intended for research use only and not for medical applications.

  • Purity of 98.70%
  • Appearance as an off-white to light yellow solid
  • Stable under recommended storage conditions
  • Soluble in DMSO at 25 mg/mL
  • Store powder at -20°C for 3 years, or 4°C for 2 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.