Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(4-Hydroxyphenyl)-1-propanol | 10210-17-0 | 152.19 | 100 MG
SDP

Catalog No. 5000415011
Encompass_Preferred

3-(4-Hydroxyphenyl)-1-propanol, also known as Dihydro-p-coumaryl alcohol, is a metabolite that can be isolated from P. radiate callus cultures and is associated with CAD activity.

  • Isolated from P. radiate callus cultures
  • Associated with CAD activity
  • Available as an off-white to light yellow solid
  • Purity of 98.70%
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 25 mg/mL

Catalog No. 50-004-15011 Supplier Medchemexpress LLC Supplier No. HYN1753100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(4-Hydroxyphenyl)-1-propanol, also known as Dihydro-p-coumaryl alcohol, is a metabolite that can be isolated from P. radiate callus cultures and is associated with CAD activity.

  • Isolated from P. radiate callus cultures
  • Associated with CAD activity
  • Available as an off-white to light yellow solid
  • Purity of 98.70%
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 25 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.