Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(4-Hydroxyphenyl)-1-propanol (Dihydro-p-coumaryl alcohol) | 10210-17-0 | 152.19 | 500 MG
SDP

Catalog No. 5000415777
Encompass_Preferred

3-(4-Hydroxyphenyl)-1-propanol (Dihydro-p-coumaryl alcohol) is a metabolite that can be isolated from P. radiata callus cultures and is associated with CAD activity. It is for research use only.

  • Purity: 98.70%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 6 months, -20°C for 1 month.
  • Solubility (in vitro): 25 mg/mL (164.27 mM) in DMSO (with ultrasonic and warming and heat to 60°C).
  • Solubility (in vivo) Protocol 1: ≥ 1 mg/mL (6.57 mM) in 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline (clear solution).
  • Solubility (in vivo) Protocol 2: 1 mg/mL (6.57 mM) in 10% DMSO, 90% (20% SBE-β-CD in Saline) (suspended solution; requires ultrasonic).
  • Solubility (in vivo) Protocol 3: ≥ 1 mg/mL (6.57 mM) in 10% DMSO, 90% Corn Oil (clear solution).

Catalog No. 50-004-15777 Supplier Medchemexpress LLC Supplier No. HYN1753500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(4-Hydroxyphenyl)-1-propanol (Dihydro-p-coumaryl alcohol) is a metabolite that can be isolated from P. radiata callus cultures and is associated with CAD activity. It is for research use only.

  • Purity: 98.70%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 6 months, -20°C for 1 month.
  • Solubility (in vitro): 25 mg/mL (164.27 mM) in DMSO (with ultrasonic and warming and heat to 60°C).
  • Solubility (in vivo) Protocol 1: ≥ 1 mg/mL (6.57 mM) in 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline (clear solution).
  • Solubility (in vivo) Protocol 2: 1 mg/mL (6.57 mM) in 10% DMSO, 90% (20% SBE-β-CD in Saline) (suspended solution; requires ultrasonic).
  • Solubility (in vivo) Protocol 3: ≥ 1 mg/mL (6.57 mM) in 10% DMSO, 90% Corn Oil (clear solution).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.