Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(4-Hydroxyphenyl)propionic acid

Catalog No. H5240625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H5240625G 25 g
H5240650G 50 g
H524065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H5240625G Supplier Sigma Aldrich Supplier No. H5240625G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(4-Hydroxyphenyl)propionic acid

Specifications

CAS 501-97-3
Melting Point 129°C to 131°C (lit.)
Molecular Formula C9H10O3
Linear Formula HOC6H4CH2CH2CO2H
MDL Number MFCD00002778
Quantity 25 g
Synonym 3-(p-Hydroxyphenyl)propionic acid; Phloretic acid
Molecular Weight (g/mol) 166.17
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.