Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(4-Hydroxyphenyl)Propionic Acid N-Hydroxysuccinimide Ester

Catalog No. H12561G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12561G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12561G Supplier Sigma Aldrich Supplier No. H12561G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(4-Hydroxyphenyl)Propionic Acid N-Hydroxysuccinimide Ester

Specifications

CAS 150-39-0
Melting Point 212°C
Molecular Formula C10H18N2O7
Linear Formula HOCH2CH2N(CH2CO2H)CH2CH2N(CH2CO2H)2
MDL Number MFCD00004294
Quantity 100 g
Synonym N-Succinimidyl 3-(4-hydroxyphenyl)propionate; N-[(4-Hydroxyhydrocinnamoyl)oxy]succinimide; Bolton-Hunter reagent precursor; Rudinger reagent; SHPP
Molecular Weight (g/mol) 278.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.