Learn More
Ampco 3/4 in. to 1 in. Ratchet Adapter
SDP
Catalog No. 19174621 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
2 -1/4 in.
Catalog No. Length (English)
19-174-621 2 -1/4 in.
1 options
Catalog No. 19-174-621 Supplier Ampco Supplier No. W140RA
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion resistant ratchet adapter. Frequently purchased with Ampco ratchet wrench.
  • Frequently purchased with ratchet wrench.
  • Made in the USA.
  • HS Code: 8207.90.
  • See image for additional information regarding measurements.
  • End 1 size (a1): 3/4 in.
  • End 2 size (a2): 1 in.
  • Length: 2 and 1/4 in.
Specifications
Product Type Ratchet Adapter
Length (English) 2 -1/4 in.
Quantity 1 Ea.