Learn More
Ace Glass, Inc. Coupling, makes bead to bead coupling of 3/4 inch glass process drain pipe, 25in-lbs max torque, max temp400°F (204°C)
SDP
Catalog No. 50979943
Encompass
Catalog No. 50-979-943 Supplier Ace Glass, Inc. Supplier No. 885605