Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 3/4 in. Sanitary Flange to 3/8 in. Barb - Polycarbonate
SDP

Catalog No. 01000715 Shop All Fisherbrand Products
Catalog No. 01-000-715

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Sanitary process fitting. Smooth inside diameter surface to reduce turbulence and dead space – 2-3 Microinches.

  • Works with all Commercially Available Clamps
  • Optimized seal with TPE tubing.
  • Tested with Flexelene™ tubing to burst of tubing at 180PSI – 285PSI
  • USP Class VI Compliant
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty and Services

Replacement if not satisfied. 12 Month return Policy.

Specifications

Certifications/Compliance Complaince Certificates Available
Description Class 7 Cleanroom Manufactured
Material Polycarbonate
For Use With (Application) Suitable for use in many industries including single use pharmaceutical, biotech, beverage, diary, food and other industries where sanitary product flow is important.
Packaging Quantity Single Pack
Storage Requirements Out of direct sunlight, >20°F <100°F
Color Transparent Purple
Product Type Sanitary Flange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.